Learn More
1-(2-Hydroxy-4-methylphenyl)ethanone, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 6921-64-8 |
| Molecular Formula | C9H10O2 |
| MDL Number | MFCD00100559 |
| Quantity | 100 g |
| SMILES | CC(C1=CC=C(C)C=C1O)=O |
| Molecular Weight (g/mol) | 150.17 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-(2-Hydroxy-4-methylphenyl)ethanone |