Learn More
1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene, 97+%
Description
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Specifications
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.57 |
| Quantity | 1 g |
| Sensitivity | Air Sensitive |
| Solubility Information | Insoluble in water. |
| Optical Rotation | +103° (c=1 in Chloroform) |
| Formula Weight | 418.58 |
| Percent Purity | ≥97% |
| Chemical Name or Material | 1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.