Learn More
1,11-Undecanedicarboxylic Acid, TRC
Product Code. 30012309
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012309 5 g 5g
30003233 10 g 10g
30006199 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012309 Supplier Toronto Research Chemicals Supplier No. TRCU7876655G
 Restricted Item
Fecha estimada de envoi: 18-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 505-52-2
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.33
Synonym 1,13-Tridecanedioic Acid, Brassylic Acid, NSC 9498, α,ω-Tridecanedioic Acid, ω-Carboxylauric Acid;
IUPAC Name tridecanedioic acid
SMILES OC(=O)CCCCCCCCCCCC(=O)O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C13H24O4/c14-12(15)10-8-6-4-2-1-3-5-7-9-11-13(16)17/h1-11H2,(H,14,15)(H,16,17)
Formula Weight 244.1675
Chemical Name or Material 1,11-Undecanedicarboxylic Acid