CAS RN 956906-93-7
IUPAC Nombre:
2-(4-{1-[(quinolin-6-yl)methyl]-1H-[1,2,3]triazolo[4,5-b]pyrazin-6-yl}-1H-pyrazol-1-yl)ethan-1-ol; methanesulfonic acid
Sinónimos:
2-{4-[3-(quinolin-6-ylmethyl)-[1,2,3]triazolo[4,5-b]pyrazin-5-yl]pyrazol-1-yl}ethanol; methanesulfonic acid
Peso molecular (g/mol):
468.49
Fórmula molecular:
C20H20N8O4S
InChi Key:
HBEMHKVWZJTVOC-UHFFFAOYSA-N
SMILES:
CS(O)(=O)=O.OCCN1C=C(C=N1)C1=CN=C2N=NN(CC3=CC4=CC=CN=C4C=C3)C2=N1
1
–
2
de
2
resultados
2
PF-04217903 methanesulfonate, MedChemExpress
MedChemExpress PF-04217903 methanesulfonate is a potent ATP-competitive c-Met kinase inhibitor with Ki of 4.8 nM for human c-Met. PF-04217903 methanesulfonate shows more than 1,000-fold selectivity relative to 208 kinases. Antiangiogenic properties.