CAS RN 95635-56-6
IUPAC Nombre:
dihydrogen N-(2,6-dimethylphenyl)-2-{4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]piperazin-1-yl}acetamide dichloride
Sinónimos:
dihydrogen ranolazine dichloride
Peso molecular (g/mol):
500.46
Fórmula molecular:
C24H35Cl2N3O4
InChi Key:
RJNSNFZXAZXOFX-UHFFFAOYNA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1OCC(O)CN1CCN(CC(=O)NC2=C(C)C=CC=C2C)CC1
1
–
3
de
3
resultados
1
Diclorhidrato de ranolazina, TRC
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
3
Ranolazine dihydrochloride, MedChemExpress
MedChemExpress Ranolazine dihydrochloride (CVT 303 dihydrochloride) is an anti-angina drug that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine dihydrochloride is also a partial fatty acid oxidation inhibitor.