CAS RN 940929-33-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ SB 743921 hydrochloride
Potent kinesin spindle protein (KSP) inhibitor
2
SB-743921 hydrochloride, MedChemExpress
MedChemExpress SB-743921 hydrochloride is a potent inhibitor of the mitotic kinesin KSP (Eg5), with a Ki of 0.1 nM.