CAS RN 923596-52-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ CDK8/19i
Potent and selective CDK8 and CDK19 inhibitor; maintains pluripotency of mouse PSCs in culture