CAS RN 91826-20-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
5
de
5
resultados
1
MedChemExpress GRF (1-29) amide (rat)
MedChemExpress GRF (1-29) amide (rat) is a synthetic peptide which can stimulate the growth hormone (GH) secretion.
2
MedChemExpress GRF (1-29) amide (rat)
MedChemExpress GRF (1-29) amide (rat) is a synthetic peptide which can stimulate the growth hormone (GH) secretion.
3
MedChemExpress GRF (1-29) amide (rat)
MedChemExpress GRF (1-29) amide (rat) is a synthetic peptide which can stimulate the growth hormone (GH) secretion.
4
MedChemExpress GRF (1-29) amide (rat)
MedChemExpress GRF (1-29) amide (rat) is a synthetic peptide which can stimulate the growth hormone (GH) secretion.