CAS RN 906480-09-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Angiopep-2-Cys
Binds to LRP1 and facilitates receptor-mediated transcytosis of cargo across the BBB