CAS RN 864915-61-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress HAEGTFTSDVS
MedChemExpress HAEGTFTSDVS is the first N-terminal 1-11 residues of GLP-1 peptide.
2
MedChemExpress HAEGTFTSDVS
MedChemExpress HAEGTFTSDVS is the first N-terminal 1-11 residues of GLP-1 peptide.
3
MedChemExpress HAEGTFTSDVS
MedChemExpress HAEGTFTSDVS is the first N-terminal 1-11 residues of GLP-1 peptide.