CAS RN 85490-53-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Motilin, canine
MedChemExpress Motilin, canine is a 22-amino acid peptide. Motilin is a potent agonist for gastrointestinal smooth muscle contraction.
2
MedChemExpress Motilin, canine
MedChemExpress Motilin, canine is a 22-amino acid peptide. Motilin is a potent agonist for gastrointestinal smooth muscle contraction.
3
MedChemExpress Motilin, canine
MedChemExpress Motilin, canine is a 22-amino acid peptide. Motilin is a potent agonist for gastrointestinal smooth muscle contraction.