CAS RN 848695-25-0
IUPAC Nombre:
6-chloro-9-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]-9H-purin-2-amine
Sinónimos:
6-chloro-9-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]purin-2-amine
Peso molecular (g/mol):
318.77
Fórmula molecular:
C14H15ClN6O
InChi Key:
QULDDKSCVCJTPV-UHFFFAOYSA-N
SMILES:
COC1=C(C)C(CN2C=NC3=C(Cl)N=C(N)N=C23)=NC=C1C
1
–
1
de
1
resultados
1
BIIB021, MedChemExpress
MedChemExpress BIIB021 (CNF2024) is an orally active, fully synthetic inhibitor of HSP90 with a Ki and an EC50 of 1.7 nM and 38 nM, respectively.