CAS RN 848193-69-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(R)-Selisistat, MedChemExpress
MedChemExpress (R)-Selisistat ((R)-EX-527) is a R-enantiomer of Selisistat. Selisistat (EX-527) is a potent and selective SIRT1 inhibitor with IC50 of 98 nM.