CAS RN 74285-86-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Triptophenolide, MedChemExpress
MedChemExpress Triptophenolide is a colorless crystalline plate isolated from ethyl acetate extracts of Tripterygium wilfordii.