CAS RN 68497-62-1
IUPAC Nombre:
N-{2-[bis(propan-2-yl)amino]ethyl}-2-(2-oxopyrrolidin-1-yl)acetamide; sulfuric acid
Sinónimos:
pramiracetam
Peso molecular (g/mol):
367.46
Fórmula molecular:
C14H29N3O6S
InChi Key:
ACSROKXFXFNERX-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.CC(C)N(CCNC(=O)CN1CCCC1=O)C(C)C
1
–
1
de
1
resultados
1
Pramiracetam, MedChemExpress
MedChemExpress Pramiracetam is a nootropic drug derived from piracetam, and is more potent.