CAS RN 55778-02-4
IUPAC Nombre:
N-[(4-{[(pyridin-2-yl)amino]methyl}phenyl)methyl]pyridin-2-amine
Sinónimos:
N-({4-[(pyridin-2-ylamino)methyl]phenyl}methyl)pyridin-2-amine
Peso molecular (g/mol):
290.37
Fórmula molecular:
C18H18N4
InChi Key:
KBVFRXIGQQRMEF-UHFFFAOYSA-N
SMILES:
C(NC1=CC=CC=N1)C1=CC=C(CNC2=CC=CC=N2)C=C1
1
–
1
de
1
resultados
1
WZ811, MedChemExpress
MedChemExpress WZ811 is an orally active, highly potent competitive antagonist of CXCR4. WZ811 efficiently inhibits CXCR4/SDF-1 (or CXCL12)-mediated modulation of cAMP levels (EC50=1.2 nM) and SDF-1 induced Matrigel invasion in cells (EC50=5.2 nM).