CAS RN 519187-97-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Edonerpic maleate, MedChemExpress
MedChemExpress Edonerpic maleate is a novel neurotrophic agent which can inhibit amyloid-β peptides (Aβ).