CAS RN 51827-01-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress Xenopsin
MedChemExpress Xenopsin, a neurotensin-like octapeptide from Xenopus laevis skin. Xenopsin is an inhibitor of Tetragastrin stimulated gastric acid secretion.
2
MedChemExpress Xenopsin
MedChemExpress Xenopsin, a neurotensin-like octapeptide from Xenopus laevis skin. Xenopsin is an inhibitor of Tetragastrin stimulated gastric acid secretion.
3
MedChemExpress Xenopsin
MedChemExpress Xenopsin, a neurotensin-like octapeptide from Xenopus laevis skin. Xenopsin is an inhibitor of Tetragastrin stimulated gastric acid secretion.
4
MedChemExpress Xenopsin
MedChemExpress Xenopsin, a neurotensin-like octapeptide from Xenopus laevis skin. Xenopsin is an inhibitor of Tetragastrin stimulated gastric acid secretion.