CAS RN 51165-05-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Substance P (7-11)
MedChemExpress Substance P (7-11) is a C-terminal fragment of Substance P which can cause an increase in the intracellular calcium concentration.
2
MedChemExpress Substance P (7-11)
MedChemExpress Substance P (7-11) is a C-terminal fragment of Substance P which can cause an increase in the intracellular calcium concentration.
3
MedChemExpress Substance P (7-11)
MedChemExpress Substance P (7-11) is a C-terminal fragment of Substance P which can cause an increase in the intracellular calcium concentration.