CAS RN 480998-12-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MC-DOXHZN hydrochloride, MedChemExpress
MedChemExpress MC-DOXHZN ((E/Z)-Aldoxorubicin) hydrochloride is an albumin-binding prodrug of Doxorubicin (DNA topoisomerase II inhibitor), with acid-sensitive properties.