CAS RN 461054-93-3
IUPAC Nombre:
tert-butyl 3-[14-methoxy-2-(2-methylpropyl)-4,7-dioxo-3,6,17-triazatetracyclo[8.7.0.0³,⁸.0¹¹,¹⁶]heptadeca-1(10),11,13,15-tetraen-5-yl]propanoate
Sinónimos:
tert-butyl 3-[14-methoxy-2-(2-methylpropyl)-4,7-dioxo-3,6,17-triazatetracyclo[8.7.0.0³,⁸.0¹¹,¹⁶]heptadeca-1(10),11,13,15-tetraen-5-yl]propanoate
Peso molecular (g/mol):
469.58
Fórmula molecular:
C26H35N3O5
InChi Key:
NXNRAECHCJZNRF-UHFFFAOYNA-N
SMILES:
COC1=CC=C2C(NC3=C2CC2N(C3CC(C)C)C(=O)C(CCC(=O)OC(C)(C)C)NC2=O)=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ Ko 143
CAS: 461054-93-3 Fórmula molecular: C26H35N3O5
2
Ko 143, MedChemExpress
MedChemExpress Ko 143 is a potent and selective ATP-binding cassette subfamily G member 2 (ABCG2/BCRP) inhibitor. Ko 143 displays >200-fold selectivity over P-gp and MRP-1 transporters.