CAS RN 4336-70-3
IUPAC Nombre:
(cyanomethyl)triphenylphosphanium chloride
Sinónimos:
(cyanomethyl)triphenylphosphanium chloride
Peso molecular (g/mol):
337.79
Fórmula molecular:
C20H17ClNP
InChi Key:
ARPLQAMUUDIHIT-UHFFFAOYSA-M
SMILES:
[Cl-].N#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
de
1
resultados
1
Cloruro de (cianometil)trifenilfosfonio, +98 %, Thermo Scientific Chemicals
CAS: 4336-70-3 Fórmula molecular: C20H17ClNP Peso molecular (g/mol): 337.79 Número MDL: MFCD00031672 Clave InChI: ARPLQAMUUDIHIT-UHFFFAOYSA-M Sinónimo: cyanomethyl triphenylphosphonium chloride,phosphonium, cyanomethyl triphenyl-, chloride,cyanomethyl triphenylphosphanium chloride,cyanomethyl triphenyl phosphanium chloride,cyanomethyltriphenylphosphonium chloride,acmc-1ctgb,wln: nc1pr&r&r &g,cyanomethyl triphenylphosphoniumchloride,cyanomethyl triphenyl phosphonium chloride PubChem CID: 197040 Nombre IUPAC: (cyanomethyl)triphenylphosphanium chloride SMILES: [Cl-].N#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1