CAS RN 406205-74-1
IUPAC Nombre:
3-[2-cyano-3-(trifluoromethyl)phenoxy]phenyl 4,4,4-trifluorobutane-1-sulfonate
Sinónimos:
3-[2-cyano-3-(trifluoromethyl)phenoxy]phenyl 4,4,4-trifluorobutane-1-sulfonate
Peso molecular (g/mol):
453.36
Fórmula molecular:
C18H13F6NO4S
InChi Key:
LWUSZIVDPJPVBW-UHFFFAOYSA-N
SMILES:
FC(F)(F)CCCS(=O)(=O)OC1=CC=CC(OC2=CC=CC(=C2C#N)C(F)(F)F)=C1
1
–
1
de
1
resultados
1
Bay 59-3074, MedChemExpress
MedChemExpress Bay 59-3074 is a selective cannabinoid CB1/CB2 receptor partial agonist with Ki values of 48.3 and 45.5 nM at human CB1 and CB2 receptors, respectively. Bay 59-3074 has analgesic properties.