CAS RN 377090-84-1
IUPAC Nombre:
3-[(1H-imidazol-5-yl)methylidene]-5-methoxy-2,3-dihydro-1H-indol-2-one
Sinónimos:
3-(3H-imidazol-4-ylmethylidene)-5-methoxy-1H-indol-2-one
Peso molecular (g/mol):
241.25
Fórmula molecular:
C13H11N3O2
InChi Key:
QNUKRWAIZMBVCU-UHFFFAOYSA-N
SMILES:
COC1=CC2=C(NC(=O)C2=CC2=CN=CN2)C=C1
1
–
2
de
2
resultados
1
SU9516, MedChemExpress
MedChemExpress SU9516 is a potent CDK2 inhibitor, with an IC50 of 22 nM, and also shows inhibitory effects on CDK1 and CDK4, with IC50s of 40, 200 nM, respectively.
2