CAS RN 35899-54-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SIBA, MedChemExpress
MedChemExpress SIBA (5'-Isobutylthioadenosine), a synthetic analogue of SAH (HY-19528), acts as an inhibitor of S-adenosylmethionine-mediated transmethylation. SIBA can interfere with a variety of enzymatic activities in vitro, such as SAH hydrolase, methylthioadenosine phosphorylase and cAMP phosphodiesterase. SIBA reversibly blocks the multiplication of herpes simplex type 1 virus (HSV).