CAS RN 342652-67-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MRK-016, MedChemExpress
MedChemExpress MRK-016 is a selective, orally bioavailable inverse agonist of GABAA α5 receptor, with an EC50 of 3 nM for GABAA α5, and Kis of 0.83, 0.85, 0.77 and 1.4 nM for human GABAA α1β3γ2, GABAA α2β3γ2, GABAA α3β3γ2, and GABAA α5β3γ2, respectively; MRK-016 also readily penetrates the CNS.