CAS RN 3023933-33-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ SCOTfluor 510 Fmoc-Dapa-OH
Fmoc protected fluorescent amino acid for use in solid-phase peptide synthesis to prepare PAINT imaging probes