CAS RN 2925481-88-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ BAY 805
Potent and selective inhibitor of USP21; also NF-κB activator