CAS RN 288250-47-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Y-320, MedChemExpress
MedChemExpress Y-320 is a new phenylpyrazoleanilide immunomodulator; inhibits IL-17 production by CD4 T cells stimulated with IL-15 with IC50 values of 20 to 60 nM.