CAS RN 2861905-57-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ ICG-d7
Near-infrared fluorescent dye; partially deuterated form of Indocyanine Green (Cat. No. 7510)