CAS RN 285983-48-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
Tocris Bioscience™ BIRB 796
High affinity and selective p38 kinase inhibitor
2
Doramapimod, TRC
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
3
4
Doramapimod, MedChemExpress
MedChemExpress Doramapimod (BIRB 796) is an orally active, highly potent p38 MAPK inhibitor, which has an IC50 for p38α=38 nM, for p38β=65 nM, for p38γ=200 nM, and for p38δ=520 nM. Doramapimod has picomolar affinity for p38 kinase (Kd=0.1 nM). Doramapimod also inhibits B-Raf with an IC50 of 83 nM.