CAS RN 2826-26-8
IUPAC Nombre:
2-[(4-methoxyphenyl)methylidene]propanedinitrile
Sinónimos:
2-[(4-methoxyphenyl)methylidene]propanedinitrile
Peso molecular (g/mol):
184.2
Fórmula molecular:
C11H8N2O
InChi Key:
UOHFCPXBKJPCAD-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C(C#N)C#N)C=C1
1
–
1
de
1
resultados
1
Tyrphostin A1, MedChemExpress
MedChemExpress Tyrphostin A1(AG9) inhibits CD40L-stimulated IL-12 production in macrophage cultures and antigen-induced generation of Th1 cells.