CAS RN 2624313-13-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SGC3027, MedChemExpress
MedChemExpress SGC3027 is a histone methyltransferase inhibitor. SGC3027 is the first potent, selective and cell active chemical probe for PRMT7.