CAS RN 243966-09-8
IUPAC Nombre:
2-(carboxyformamido)-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carboxylic acid hydrochloride
Sinónimos:
2-(carboxyformamido)-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carboxylic acid hydrochloride
Peso molecular (g/mol):
306.72
Fórmula molecular:
C10H11ClN2O5S
InChi Key:
LQGCAMWQDSYOAY-UHFFFAOYSA-N
SMILES:
Cl.OC(=O)C(=O)NC1=C(C(O)=O)C2=C(CNCC2)S1
1
–
1
de
1
resultados
1
TCS 401, MedChemExpress
MedChemExpress TCS 401 is a selective inhibitor of protein tyrosine phosphatase 1B (PTP1B).