CAS RN 2417507-09-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ SMCy 5.5
Fluorogenic fluorescent probe for imaging lipid droplets; suitable for use with two-photon microscopy