CAS RN 2378-86-1
IUPAC Nombre:
[(4-methylphenyl)methyl]triphenylphosphanium bromide
Sinónimos:
[(4-methylphenyl)methyl]triphenylphosphanium bromide
Peso molecular (g/mol):
447.36
Fórmula molecular:
C26H24BrP
InChi Key:
CEEKCHGNKYVTTM-UHFFFAOYSA-M
SMILES:
[Br-].CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
1
–
1
de
1
resultados
1
Bromuro de (4-metilbencílico)trifenilfosfonio, + 98 %, Thermo Scientific Chemicals
CAS: 2378-86-1 Fórmula molecular: C26H24BrP Peso molecular (g/mol): 447.36 Número MDL: MFCD00031580 Clave InChI: CEEKCHGNKYVTTM-UHFFFAOYSA-M Sinónimo: 4-methylbenzyl triphenylphosphonium bromide,4-methylphenyl methyl triphenylphosphanium bromide,pubchem2184,acmc-1cfpa,4-methylphenyl methyl-triphenyl-phosphanium bromide,4-methylbenzyl triphenylphosphoniumbromide,4-methylphenyl methyl-triphenylphosphonium bromide,p-methylbenzyl-triphenylphosphomiumbromide,phosphonium, 4-methylphenyl methyl triphenyl-, bromide 1:1 PubChem CID: 11797662 SMILES: [Br-].CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1