CAS RN 2364489-81-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ FMF-04-159-2
Potent covalent CDK14 and CDK16 inhibitor; also inhibits other TAIRE kinase family members