CAS RN 2310134-98-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LDN193189 Tetrahydrochloride, MedChemExpress
MedChemExpress LDN193189 Tetrahydrochloride is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM).