CAS RN 2245946-45-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Janelia Fluor 549, free acid
Yellow dye (Reactive group: free acid). Applications: super resolution microscopy e.g. dSTORM & STED, confocal, flow and live cell imaging