CAS RN 2236-01-3
IUPAC Nombre:
(2-oxopropyl)triphenylphosphanium bromide
Sinónimos:
(2-oxopropyl)triphenylphosphanium bromide
Peso molecular (g/mol):
399.27
Fórmula molecular:
C21H20BrOP
InChi Key:
KZBWHCFTAAHIJJ-UHFFFAOYSA-M
SMILES:
[Br-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
de
1
resultados
1
Bromuro de acetoniltrifenilfosfonio, +98 %, Thermo Scientific Chemicals
CAS: 2236-01-3 Fórmula molecular: C21H20BrOP Peso molecular (g/mol): 399.27 Número MDL: MFCD00051863 Clave InChI: KZBWHCFTAAHIJJ-UHFFFAOYSA-M Sinónimo: acetonyltriphenylphosphonium bromide,acetonyl triphenylphosphonium bromide,2-oxopropyl triphenylphosphanium bromide,phosphonium, 2-oxopropyl triphenyl-, bromide,acetonyltriphenylphosphoniumbromide,acmc-1cmpe,2-oxopropyl-triphenylphosphonium bromide,2-oxopropyl triphenylphosphonium bromide,2-oxopropyl triphenyl phosphanium bromide,phosphonium, 2-oxopropyl triphenyl-,bromide 1:1 PubChem CID: 638158 Nombre IUPAC: 2-oxopropil(trifenil)fosfanio; bromuro SMILES: [Br-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1