CAS RN 21888-98-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Dexetimide, MedChemExpress
MedChemExpress Dexetimide ((+)-Benzetimide) is a high-affinity muscarinic receptor antagonist and a potent and persistent anticholinergic agent used to treat neuroleptic-induced parkinsonism.