CAS RN 201422-04-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Autocamtide-2-related inhibitory peptide, myristoylated
CaM kinase II inhibitor; enhanced cell permeable derivative of Autocamtide-2-related inhibitory peptide