CAS RN 2012607-27-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
WNK463, MedChemExpress
MedChemExpress WNK463 is an orally bioavailable pan-With-No-Lysine (K) (WNK)-kinase inhibitor with IC50s of 5 nM, 1 nM, 6 nM, and 9 nM for WNK1, WNK2, WNK3, and WNK4, respectively.