CAS RN 1950634-92-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ dBET6
Potent and selective (+)-JQ1 based Degrader (PROTAC™) targeting BET bromodomains, active in vivo
2
dBET6, MedChemExpress
MedChemExpress dBET6 is a highly potent, selective and cell-permeable PROTAC connected by ligands for Cereblon and BET, with an IC50 of 14 nM, and has antitumor activity.