CAS RN 188011-69-0
IUPAC Nombre:
3-[(5-bromopyridin-2-yl)carbamoyl]propanoic acid
Sinónimos:
3-[(5-bromopyridin-2-yl)carbamoyl]propanoic acid
Peso molecular (g/mol):
273.09
Fórmula molecular:
C9H9BrN2O3
InChi Key:
XFYYQDHEDOXWGA-UHFFFAOYSA-N
SMILES:
OC(=O)CCC(=O)NC1=NC=C(Br)C=C1
1
–
1
de
1
resultados
1
Bikinin, MedChemExpress
MedChemExpress Bikinin is a non-steroidal, ATP-competitive inhibitor of plant GSK-3/Shaggy-like kinases and activates BR (brassinosteroids) signaling.