CAS RN 1820684-31-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
M443, MedChemExpress
MedChemExpress M443 is an irreversible and specific inhibitor of MRK, with an IC50<125 nM.