CAS RN 1811539-57-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Janelia Fluor™ 646, free acid
Red dye (Reactive group: free acid). Application: flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM & STED. Suitable for live cell imaging