CAS RN 1811539-32-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Janelia Fluor 549, SE
Yellow dye (Reactive group: NHS ester). Applications: super resolution microscopy e.g. dSTORM & STED, confocal, flow and live cell imaging