CAS RN 179474-81-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Prucalopride, MedChemExpress
MedChemExpress Prucalopride (R093877) is a drug acting as a selective, high affinity 5-HT4 receptor agonist(pKi=8.6/8.1 for 5-HT4a/4b); >150-fold higher affinity for 5-HT4 receptors than for other receptors.