CAS RN 174634-09-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
TAS-103 dihydrochloride, MedChemExpress
MedChemExpress TAS-103 dihydrochloride is a dual inhibitor of DNA topoisomerase I/II, used for cancer research.